missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FosB Recombinant Rabbit Monoclonal Antibody (6M9D5)

Rabbit Recombinant Monoclonal Antibody
€ 176.00 - € 436.00
Specifications
| Antigen | FosB |
|---|---|
| Clone | 6M9D5 |
| Concentration | 2.16 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
30282853
|
Invitrogen™
MA558099 |
20 μL |
€ 176.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284159
![]() |
Invitrogen™
MA558100 |
100 μL |
€ 436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Immunogen sequence: MFQAFPGDYD SGSRCSSSPS AESQYLSSVD SFGSPPTAAA SQECAGLGEM PGSFVPTVTA ITTSQDLQWL VQPTLISSMA QSQGQPLASQ PPVVDPYDMP.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.Specifica
| FosB | |
| 2.16 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| D3ZLB7, P53539 | |
| 100360880, 2354 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Fos B (NP_006723.2). | |
| Antibody |
| 6M9D5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| Fosb | |
| activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
| Fosb | |
| Primary | |
| Protein A |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto