missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosfomycin Calcium Monohydrate, Mikromol™
Description
Fosfomycin Calcium Monohydrate
Specifications
Specifications
| Chemical Name or Material | Fosfomycin Calcium Monohydrate |
| CAS | 26469-67-0 |
| Analyte or Component Names | Fosfomycin Calcium Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H5 O4 P . Ca . H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Fosfomycin calcium, Calcium (2R,3S)-(3-methyloxiran-2-yl)phosphonate monohydrate |
| InChI Formula | InChI=1S/C3H7O4P.Ca.H2O/c1-2-3(7-2)8(4,5)6;;/h2-3H,1H3,(H2,4,5,6);;1H2/q;+2;/p-2/t2-,3+;;/m0../s1 |
| SMILES | O.[Ca+2].C[C@@H]1O[C@@H]1P(=O)([O-])[O-] |
| IUPAC Name | calcium;[(2R,3S)-3-methyloxiran-2-yl]-dioxido-oxo-λ^{5}-phosphane;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?