Learn More
FOSL1 Mouse anti-Human, Clone: OTI12F9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500624
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The v-Fos oncogene was initially detected in two independent murine osteosarcoma virus isolates and an avian nephroblastoma virus. Members of the c-Fos gene family, including c-Fos, Fos B, Fra-1 and Fra-2, encode nuclear phosphoproteins that are rapidly and transiently induced by a variety of agents and function as transcriptional regulators for several genes. In contrast to c-Jun proteins, which form homo- and heterodimers that bind to specific DNA response elements, c-Fos proteins are only active as heterodimers with members of the Jun gene family. In addition, selected ATF/CREB family members can form leucine zipper dimers with Fos and Jun. Different dimers exhibit differential specificity and affinity for AP-1 and CRE sites.Specifications
| FOSL1 | |
| Monoclonal | |
| Unconjugated | |
| FOSL1 | |
| FRA, FRA1, fra-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8061 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI12F9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15407 | |
| FOSL1 | |
| Full-length protein expressed in 293T cell transfected with human FOSL1 expression vector | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.