missing translation for 'onlineSavingsMsg'
Learn More

Fostamatinib disodium hexahydrate, MedChemExpress

Product Code. 16431687
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431687

Brand: Medchem Express HY13038B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fostamatinib (R788) disodium hexahydrate is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

Fostamatinib (R788) disodium hexahydrate is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

Specifications

Chemical Name or Material Fostamatinib disodium hexahydrate
CAS 914295-16-2
Color White
Purity Grade Notes Research
Molecular Formula C23H36FN6Na2O15P
Quantity 50 mg
Synonym R788 disodium hexahydrate
Solubility Information DMSO : 31.81 mg/mL (43.43 mM; Need ultrasonic) ∣H2O : 2 mg/mL (2.73 mM; ultrasonic and warming and heat to 60°C)
SMILES COC1=CC(NC2=NC=C(C(NC3=NC(N4COP(O[Na])(O[Na])=O)=C(C=C3)OC(C)(C)C4=O)=N2)F)=CC(OC)=C1OC.O.O.O.O.O.O
Molecular Weight (g/mol) 732.51
Formula Weight 732.51
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.