Learn More
Duran™ Baffled Flask
Provide increased agitation at lower speeds for a more gentle medium aeration of sensitive cell cultures. Duran™ Four Bottom Borosilicate Glass Baffled Flask molded bottom increases aeration for better growth, especially with bacteria.
Brand: Duran™ 212834405
Additional Details : Weight : 1.70100kg
Description
- Consistent flask shapes for highly reproducible experimental conditions and results
- Borosillicate glass with high chemical and thermal shock resistance
- GL 45 thread necks
- Ehrlenmeyer
- Four bottom baffles increase aeration at higher shaking speeds