missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOX2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 321.00 - € 493.00
Specifications
| Antigen | FOX2 |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666676
|
Novus Biologicals
NBP3-05534-100ul |
100 μg |
€ 493.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633139
|
Novus Biologicals
NBP3-05534-50ul |
50 μg |
€ 321.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOX2 Polyclonal antibody specifically detects FOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FOX2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Cell Cycle and Replication | |
| Supplied as an aliquot of IgY preparation | |
| 23543 | |
| IgY | |
| IgY purified |
| Western Blot 1:1000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dJ106I20.3, Fox-1 homolog B, fox-1 homologue, FOX2Fox-2, fxh, hexaribonucleotide binding protein 2, Hexaribonucleotide-binding protein 2, HRNBP2FOX-2, RBM9RTAHNRBP2, Repressor of tamoxifen transcriptional activity, RNA binding motif protein 9, RNA binding protein fox-1 homolog 2, RNA binding protein, fox-1 homolog (C. elegans) 2, RNA-binding motif protein 9, RNA-binding protein 9 | |
| N-terminal 99 amino acids of human FOX2 expressed in and purified from E. coli | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title