missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 Mouse anti-Human, Clone: FOXA1/1241, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14810.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human FOXA1.
- High specificity
- High reactivity
Specifications
| FOXA1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined | |
| P55317 | |
| FOXA1 | |
| Recombinant protein corresponding to amino acids 467-614 of human FOXA1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| FOXA1/1241 | |
| Mouse monoclonal antibody raised against partial recombinant human FOXA1. | |
| In 10 mM PBS. | |
| HNF3A/MGC33105/TCF3A | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3169 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction