missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXA2 Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142064
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms: NP_068556.2; NP_710141.1. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
Specifications
| FOXA2 | |
| Polyclonal | |
| Biotin | |
| FOXA2 | |
| FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 25099, 3170 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P32182, Q9Y261 | |
| FOXA2 | |
| Peptide with sequence C-GVYSRPIMNSS (aa 447-457). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction