missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB1 Antibody (PCRP-FOXB1-1B7), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14021MFV610
This item is not returnable.
View return policy
Description
FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for Flow Cytometry
Specifications
| FOXB1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 27023 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| PCRP-FOXB1-1B7 | |
| Flow Cytometry | |
| FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5 | |
| Recombinant full-length human FOXB1protein (Uniprot: Q99853) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction