missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxC2 Antibody (OTI3E3), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70752R
This item is not returnable.
View return policy
Description
FoxC2 Monoclonal specifically detects FoxC2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FoxC2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FOXC2 | |
| Recombinant protein fragment corresponding to amino acids 1-280 of human FoxC2 (NP_005242) produced in E.coli. | |
| 0.1 mL | |
| 2303.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3E3 | |
| Western Blot | |
| FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction