Learn More
FOXC2 Mouse anti-Human, Clone: OTI2G5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807691
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.Specifications
| FOXC2 | |
| Monoclonal | |
| Unconjugated | |
| FOXC2 | |
| FKHL14, LD, MFH-1, MFH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2303 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2G5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99958 | |
| FOXC2 | |
| Human recombit protein fragment corresponding to amino acids 1-280 of human FOXC2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.