missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161534 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161534 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161534 Supplier Invitrogen™ Supplier No. PA5113676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GMGRSYAPYH HQPAAPKDLV KPPYSYIALI TMAIQNAPEK KITLNGIYQF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 100%; Rat: 100%.

Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXC2
Gene Accession No. Q61850
Gene Alias BF-3; brain factor 3; Fkh14; Fkhl14; forkhead box C2; forkhead box C2 (MFH-1 mesenchyme forkhead 1); forkhead box C2 (MFH-1, mesenchyme forkhead 1); forkhead box protein C2; forkhead homolog 14; forkhead, Drosophila, homolog-like 14; forkhead-related protein FKHL14; FOXC2; Hfhbf3; HFH-BF-3; LD; mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH1; MFH-1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; Transcription factor FKH-14
Gene Symbols FOXC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Mouse Foxc2 (aa 51-100).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14234
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.