missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1270
This item is not returnable.
View return policy
Description
FOXD2 Polyclonal specifically detects FOXD2 in Human, Mouse, Rat samples. It is validated for Peptide ELISA.
Specifications
| FOXD2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FKHL17forkhead, drosophila, homolog-like 17, forkhead box D2, forkhead-like 17, forkhead-related activator 9, Forkhead-related protein FKHL17, Forkhead-related transcription factor 9, FREAC-9, FREAC9forkhead box protein D2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2306 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:8000 | |
| NP_004465. | |
| FOXD2 | |
| Peptide with sequence FASKVAGLSGCHF corresponding to C-Terminus according to NP_004465.3. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction