missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98870-100ul
This item is not returnable.
View return policy
Description
FoxF1 Polyclonal antibody specifically detects FoxF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FoxF1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human FoxF1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2294 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction