missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583039
This item is not returnable.
View return policy
Description
Immunogen sequence: GFRRKCQALK PMYSMMNGLG FNHLPDTYGF QGSAGGLSCP PNSLALEGGL GMMNGHLPGN VDGMALPSHS VPHLPSNGGH SYMGG Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq, Jul 2008].
Specifications
| FOXF1 | |
| Polyclonal | |
| Unconjugated | |
| Foxf1 | |
| ACDMPV; AI450827; FKHL5; forkhead box F1; forkhead box F1a; forkhead box protein F1; forkhead box protein F1; LOW QUALITY PROTEIN: forkhead box protein F1; Forkhead, drosophila, homolog-like 5; forkhead-related activator 1; Forkhead-related protein FKHL5; forkhead-related transcription factor 1; Foxf1; Foxf1a; FREAC1; Freac-1; Hepatocyte nuclear factor 3 forkhead homolog 8; Hfh8; HFH-8; HNF-3/forkhead homolog 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12946 | |
| Foxf1 | |
| Recombinant protein corresponding to Human FOXF1. Recombinant protein control fragment (Product #RP-100730). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction