missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxH1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41196AF700
This item is not returnable.
View return policy
Description
FoxH1 Polyclonal antibody specifically detects FoxH1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| FoxH1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 700 | |
| Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer | |
| Antibody was raised against a 19 amino acid peptide near the amino terminus of human FOXH1. The immunogen is located within the first 50 amino acids of FOXH1. Amino Acid Squence: CQPPKRRKKRYLRHDKPPYT | |
| 0.1 mL | |
| Stem Cell Markers | |
| 8928 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction