missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI1 Monoclonal Antibody (OTI2G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800146
This item is not returnable.
View return policy
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| FOXI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12951 | |
| FOXI1 | |
| Human recombit protein fragment corresponding to amino acids 111-350 of human FOXI1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G6 | |
| Unconjugated | |
| FOXI1 | |
| FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2299 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction