missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxJ1/HFH4 Antibody (407003) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3619JF635
This item is not returnable.
View return policy
Description
FoxJ1/HFH4 Monoclonal antibody specifically detects FoxJ1/HFH4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| FoxJ1/HFH4 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202 | |
| E. coli-derived recombinant human FoxJ1, Lys306-Leu421, Accession # Q92949.3 | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 2302 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 407003 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction