missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXJ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568947
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 100%; Rabbit: 100%; Rat: 93%.
Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.
Specifications
| FOXJ1 | |
| Polyclonal | |
| Unconjugated | |
| FOXJ1 | |
| FKHL13; FKHL-13; fork head homologue 4; forkhead box J1; forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; Foxj1; Hepatocyte nuclear factor 3 forkhead homolog 4; Hfh4; HFH-4; HNF-3/forkhead homolog 4; MGC35202 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q61660 | |
| FOXJ1 | |
| synthetic peptide corresponding to a region of mouse FOXJ1 (aa 201-250). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction