missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxJ3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71862AF350
This item is not returnable.
View return policy
Description
FoxJ3 Polyclonal antibody specifically detects FoxJ3 in Human samples. It is validated for Western Blot
Specifications
| FoxJ3 | |
| Polyclonal | |
| Western Blot | |
| forkhead box J3, forkhead box protein J3, KIAA1041, MGC165036, MGC176686 | |
| The immunogen for this product maps to a region between residue 525 and 575 of human Forkhead Box J3 using the numbering given in entry NP_055762.3 (GeneID 22887). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 22887 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction