missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-88146
This item is not returnable.
View return policy
Description
FoxK1 Polyclonal specifically detects FoxK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| FoxK1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| FLJ14977, forkhead box K1, forkhead box protein K1, FOXK1L, IMAGE:5164497, MNF, Myocyte nuclear factor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 221937 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-5 mcg/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_001032242.1 | |
| FOXK1 | |
| The immunogen recognized by this antibody maps to a region between residue 683 and 733 of human forkhead box K1 using the numbering given in entry NP_001032242.1 (GeneID 221937). | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Bovine, Horse, Guinea pig_10141, Pig, Panda, Rhesus Monkey, Gorilla, Chimpanzee, and Thirteen-lined ground squirrel. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction