missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Antibody (FOXL2/7989R), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20964AF532
This item is not returnable.
View return policy
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FOXL2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| Recombinant full-length human FOXL2 protein | |
| 0.1 mL | |
| Epigenetics | |
| 668 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FOXL2/7989R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction