missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN2 Monoclonal Antibody (OTI7D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812453S
This item is not returnable.
View return policy
Description
FOXN2 Monoclonal antibody specifically detects FOXN2 in Human samples. It is validated for Western BlotSpecifications
| FOXN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5JXB2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 389898 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7D12 | |
| Unconjugated | |
| UBE2NL | |
| 3230402J05Rik; 6030465J18Rik; Fkh19; forkhead box N1; forkhead box N2; forkhead box protein N2; forkhead homolog 19; Foxn1; FOXN2; HTLF; Human T-cell leukemia virus enhancer factor; RGD1562291 | |
| Human recombinant protein fragment corresponding to amino acids 1-100 of human UBE2NL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction