missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR Antibody (CL13278), Novus Biologicals™
Mouse Monoclonal Antibody
€ 350.00 - € 526.00
Specifications
| Antigen | FoxO1/FKHR |
|---|---|
| Clone | CL13278 |
| Dilution | Western Blot 1 μg/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697214
|
Novus Biologicals
NBP3-18557-100ul |
100 μg |
€ 526.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663575
|
Novus Biologicals
NBP3-18557-25ul |
25 μg |
€ 350.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FoxO1/FKHR Monoclonal antibody specifically detects FoxO1/FKHR in Human samples. It is validated for Western BlotSpecifications
| FoxO1/FKHR | |
| Western Blot 1 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| This antibody was developed against a sythetic peptide for Q12778. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL13278 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
| PBS, pH 7.2, 40% glycerol | |
| 2308 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title