missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2312C
This item is not returnable.
View return policy
Description
FoxO1/FKHR Polyclonal antibody specifically detects FoxO1/FKHR in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| FoxO1/FKHR | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| The immunogen recognized by this antibody maps to a region between residues 600 and the C-terminus (residue 652) of human Forkhead Box O1a using the numbering given in Swiss-Prot entry Q12778 (GeneID 2308). | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
| 2308 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction