Learn More
FOXO1 Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810711S
Encompass_Preferred
 View more versions of this product
Product Code. 17584124
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
FOXO1 Monoclonal antibody specifically detects FOXO1 in Human samples. It is validated for Western Blot
Specifications
Specifications
FOXO1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8N945
Mouse
Affinity chromatography
RUO
153768
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2C6
Unconjugated
PRELID2
Afxh; AI876417; FKH1; FKHR; Fkhr1; forkhead box O1; forkhead box O1 a; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box protein O1; forkhead box protein O1; LOW QUALITY PROTEIN: forkhead box protein O1; forkhead box protein O1A; Forkhead box protein O1-A; Forkhead in rhabdomyosarcoma; forkhead protein 1; forkhead protein FKHR; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; fox family transcription factor FoxO1a.1; FOX01A; foxhead box protein O1-A; foxo1; FOXO1A; FoxO1a.1; member of FKHR-subclass forkhead/winged helix family; zgc:153388
Full length human recombinant protein of human PRELID2 produced in E.coli.
30 μL