missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB10970.100uL
This item is not returnable.
View return policy
Specifications
| FOXO1 | |
| Polyclonal | |
| Unconjugated | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| FKH1/FKHR/FOXO1A | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| forkhead box O1 | |
| FOXO1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S319 of human FOXO1. | |
| RUO | |
| 2308 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction