missing translation for 'onlineSavingsMsg'
Learn More

FOXO4 Recombinant Rabbit Monoclonal Antibody (23GB1470), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

€ 207.00 - € 562.00

Specifications

Antigen FOXO4
Clone 23GB1470
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30087654
GREENER_CHOICE
View Documents
Invitrogen
MA553994
100 μL
€ 562.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30087707
View Documents
Invitrogen
MA553995
20 μL
€ 207.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FOXO4 Recombinant Monoclonal antibody specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

FOXO4
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P98177, Q9WVH3
302415, 4303, 54601
IgG
Affinity Chromatography
Antibody
23GB1470
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201
A synthesized peptide derived from human FoxO4 (450-505AA).
Primary
-20°C
FOXO4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.