missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXO6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106411
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FOXO6.
FOXO6 is a transcriptional activator.
Specifications
| FOXO6 | |
| Polyclonal | |
| Unconjugated | |
| FOXO6 | |
| forkhead box O6; forkhead box protein O6; Foxo6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100132074, 313558, 329934 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A8MYZ6, Q70KY4 | |
| FOXO6 | |
| A synthesized peptide derived from human FOXO6(Accession A8MYZ6), corresponding to amino acid residues R209-A259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction