missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP1 Antibody (JC12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65125MFV500
This item is not returnable.
View return policy
Description
FoxP1 Monoclonal antibody specifically detects FoxP1 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| FoxP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| JC12 | |
| Flow Cytometry | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4 | |
| Human FOXP1 [Uniprot# Q9H334] | |
| 0.1 mL | |
| Cancer | |
| 27086 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction