missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP1 Antibody (OTI4F1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71831AF350
This item is not returnable.
View return policy
Description
FoxP1 Monoclonal specifically detects FoxP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| FoxP1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FOXP1 | |
| Full length recombinant protein of human FoxP1 (NP_116071) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4F1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4 | |
| Mouse | |
| 0.1 mL | |
| 27086 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction