missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99918PE
This item is not returnable.
View return policy
Description
GSTM2 Polyclonal antibody specifically detects GSTM2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| GSTM2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2 | |
| Produced in rabbits immunized with purified, recombinant Human GSTM2 (Accession#: P28161; Met 1-Lys 218) | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 2946 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction