missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP2 Antibody (CL5312), Novus Biologicals™
Mouse Monoclonal Antibody
€ 379.00 - € 500.00
Specifications
| Antigen | FoxP2 |
|---|---|
| Clone | CL5312 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277991
|
Novus Biologicals
NBP2-61413 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664218
|
Novus Biologicals
NBP2-61413-25ul |
25 μL |
€ 379.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FoxP2 Monoclonal specifically detects FoxP2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FoxP2 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein | |
| FOXP2 | |
| IgG1 | |
| Protein A purified |
| CL5312 | |
| Monoclonal | |
| Purified | |
| Human, Mouse, Rat | |
| 93986 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AQQLVFQQQLLQMQQLQQQQHLLSLQRQGLISIPPGQAALPVQSLPQAGLSPAEIQQLWKEVTGVHSMEDNGIKHGGLDLTTNNSSSTTSSNTSKASPPITHHS | |
| Primary | |
| Specificity of human FoxP2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title