missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP2 Antibody (RAB-S249), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20155AF594
This item is not returnable.
View return policy
Description
FoxP2 Monoclonal antibody specifically detects FoxP2 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| FoxP2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP2 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| RAB-S249 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 93986 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction