missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP2 Mouse anti-Human, Clone:2G11B8, Abnova™
Mouse monoclonal antibody raised against recombinant human FOXP2.
Brand: Abnova MAB17175.100ug
This item is not returnable.
View return policy
Description
This gene encodes an evolutionarily conserved transcription factor expressed in fetal and adult brain. This transcription factor is a member of the forkhead/winged-helix (FOX) family of transcription factors, and contains a FOX DNA-binding domain and a large polyglutamine tract. Members of the FOX family of transcription factors are regulators of embryogenesis. The product of this gene is thought to be required for proper development of speech and language regions of the brain during embryogenesis. Although a point mutation in this gene has been associated with the KE pedigree segregating developmental verbal dyspraxia, no association between mutations in this gene and another speech disorder, autism, has been found. Multiple alternative transcripts encoding different isoforms have been identified. [provided by RefSeq]Specifications
| FOXP2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| FOXP2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 2G11B8 | |
| forkhead box P2 | |
| In PBS (0.05% sodium azide) | |
| CAGH44, DKFZp686H1726, SPCH1, TNRC10 | |
| Recombinant protein corresponding to amino acid 641-740 of human FOXP2 from E. coli. | |
| 93986 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction