missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXP3 Monoclonal Antibody (FJK-16s), Brilliant Violet™ 605, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
€ 175.00 - € 371.00
Specifications
| Antigen | FOXP3 |
|---|---|
| Clone | FJK-16s |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088202
|
Invitrogen™
406-5773-80 |
25 μg |
€ 175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30087860
![]() |
Invitrogen™
406-5773-82 |
100 μg |
€ 371.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXP3 Monoclonal antibody specifically detects FOXP3 in Bovine, Canine, Feline, Mouse, Porcine, Rat samples. It is validated for Flow CytometrySpecifications
| FOXP3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| FJK-16s | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| Rat | |
| Bovine, Canine, Feline, Mouse, Pig, Rat | |
| Q99JB6 | |
| 100037405, 20371, 317382, 444998, 491876, 506053 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Foxp3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title