missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Foxp3 Mouse, APC, Clone: 3G3, enQuireBio™
Mouse Monoclonal Antibody
€ 407.40 - € 526.05
Specifications
| Antigen | Foxp3 |
|---|---|
| Clone | 3G3 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15952398
|
EnquireBio
QAB71-APC-25ug |
25 μg |
€ 407.40
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15942398
|
EnquireBio
QAB71-APC-100ug |
100 μg |
€ 526.05
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Foxp3 | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH7.2 | |
| 20371 | |
| IgG1 κ | |
| Protein A/G Purified |
| 3G3 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_001186276 | |
| Foxp3 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title