missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Foxp3 Mouse, PE, Clone: 3G3, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB71-PE-25ug
This item is not returnable.
View return policy
Specifications
| Foxp3 | |
| Monoclonal | |
| 0.2mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH7.2 | |
| Foxp3 | |
| Protein A/G Purified | |
| RUO | |
| 20371 | |
| IgG1 κ |
| Flow Cytometry | |
| 3G3 | |
| PE | |
| NP_001186276 | |
| Mouse | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction