Learn More
Invitrogen™ FOXP3 Polyclonal Antibody

Description
PA1-806 detects FOXP3 from human samples. PA1-806 has been successfully used in Western blot and ChIP procedures. By Western blot, PA1-806 detects a ~50 kDa band representing FOXP3 from Hela cells. The PA1-806 immunogen is a synthetic peptide corresponding to residues W(406) T V D E L E F R K K R S Q R(420) of human FOXP3. PA1-806 can be used with blocking peptide PEP-297.
Specifications
Specifications
| Antigen | FOXP3 |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | Foxp3 |
| Gene Accession No. | Q9BZS1 |
| Gene Alias | AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID |
| Gene Symbols | Foxp3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.