missing translation for 'onlineSavingsMsg'
Learn More

Foxp3 Rat, APC, Clone: MF23, enQuireBio™

Rat Monoclonal Antibody

€ 248.85 - € 512.40

Specifications

Antigen Foxp3
Clone MF23
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15991748
View Documents
EnquireBio
QAB27-APC-25ug
25 μg
€ 248.85
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15981748
View Documents
EnquireBio
QAB27-APC-100ug
100 μg
€ 512.40
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Foxp3
0.2mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
20371
IgG2b
Protein A/G Purified
MF23
Flow Cytometry
APC
RUO
NP_001186276
Foxp3
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.