missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FoxP4 Antibody (PCRP-FOXP4-1G7) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27048AF700
This item is not returnable.
View return policy
Description
FoxP4 Monoclonal antibody specifically detects FoxP4 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| FoxP4 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| FKHLA, FLJ40908, FLJ44184, fork head-related protein like A, Fork head-related protein-like A, forkhead box P4, forkhead box protein P4, hFKHLA, winged-helix repressor FOXP4 | |
| Recombinant fragment (around aa302-541) of human FoxP4 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| PCRP-FOXP4-1G7 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 116113 | |
| Store at 4°C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction