missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP4 Antibody (RAB-S40), DyLight 594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20153DL594
This item is not returnable.
View return policy
Description
FoxP4 Monoclonal antibody specifically detects FoxP4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| FoxP4 | |
| Monoclonal | |
| DyLight 594 | |
| FKHLA, FLJ40908, FLJ44184, fork head-related protein like A, Fork head-related protein-like A, forkhead box P4, forkhead box protein P4, hFKHLA, winged-helix repressor FOXP4 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP4 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| RAB-S40 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 116113 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction