missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXQ1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1283
This item is not returnable.
View return policy
Description
FOXQ1 Polyclonal specifically detects FOXQ1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| FOXQ1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_150285. | |
| FOXQ1 | |
| Peptide with sequence KLEVFVPRAAHGD-C corresponding to N-Terminus according to NP_150285.3. | |
| 0.1 mg | |
| Cancer | |
| 94234 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction