missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599625
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FoxR1.
The forkhead-box (FOX) genes comprise a superfamily of at least 43 members that express proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 amino acids. FOX transcription factors play roles in determining cell fate during early development. FOXR1 (forkhead box protein R1), also known as FOXN5 (forkhead box protein N5) or DLNB13, is a 292 amino acid nuclear protein that belongs to the FOX family and contains one forkhead DNA-binding domain. Through chromosomal aberrations such as retroviral integration, gene amplification or translocation, FOXR1 may be involved the development of certain invasive carcinomas.
Specifications
| FOXR1 | |
| Polyclonal | |
| Unconjugated | |
| Foxr1 | |
| DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 283150, 315601, 382074 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UTB7, Q6PIV2 | |
| Foxr1 | |
| A synthesized peptide derived from human FOXR1(Accession Q6PIV2), corresponding to amino acid residues F231-S281. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction