missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Antibody (005), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90065AF700
This item is not returnable.
View return policy
Description
VSIG4 Monoclonal specifically detects VSIG4 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| VSIG4 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11326 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 5 | |
| ELISA, Sandwich ELISA Detection | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human VSIG4 (Q9Y279-1; Met 1-Pro 283). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction