missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXR2 Polyclonal Antibody

FOXR2 Polyclonal Antibody
Brand: Invitrogen PA598532
This item is not returnable.
View return policy
Description
FOXR2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| FOXR2 | |
| Polyclonal | |
| Unconjugated | |
| FOXR2 | |
| EG436240; Forkhead box protein N6; forkhead box protein R2; forkhead box R2; Foxn6; Foxr2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 139628 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6PJQ5 | |
| FOXR2 | |
| Recombinant Human Forkhead box protein R2 protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction