missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FPGS Recombinant Rabbit Monoclonal Antibody (7E2H1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555448
This item is not returnable.
View return policy
Description
Immunogen Sequence: NADQQNFTVT LDQVLLRCLE HQQHWNHLDE EQASPDLWSA PSPEPGGSAS LLLAPHPPHT CSASSLVFSC ISHALQWISQ GRDPIFQPPS PPKGLLTHPV AHSGASILRE AAAIHVLVTG SLHLVGGVLK LLEPALSQ.
Folylpoly-gamma-glutamate synthetase (FPGS) catalyzes the addition of several equivalents of glutamic acid to the gamma-carboxyl group in the side chain of folate cofactors and analogs. Folylpoly-gamma-glutamate synthetase has three functions in folate homeostasis in mammals: polyglutamation prevents efflux of folate cofactors from the cell, it increases the binding of folate cofactors to some of the enzymes of folate interconversion and biosynthesis, and it appears to allow the accumulation of folates in the mitochondria that are required for glycine synthesis. Folylpoly-gamma-glutamate synthetase is widely expressed in human tumors and is tightly linked either to proliferation or to a lack of differentiation. The cytotoxicity of both thymidylate synthase and purine inhibitors requires continued inhibition of target for greater than one generation time, so that the integrative function of FPGS adds considerably to the efficiency of folate antimetabolites.
Specifications
| FPGS | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q05932 | |
| FPGS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-587 of human FPGS. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7E2H1 | |
| Unconjugated | |
| FPGS | |
| AA408187; folylpoly-gamma-glutamate synthetase; folylpolyglutamate synthase; folylpolyglutamate synthase, mitochondrial; folylpolyglutamyl synthetase; FPGS; Tetrahydrofolate synthase; tetrahydrofolylpolyglutamate synthase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2356, 687266 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction