missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPR1 Antibody (350418) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3744JF635
This item is not returnable.
View return policy
Description
FPR1 Monoclonal antibody specifically detects FPR1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| FPR1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| fMet-Leu-Phe receptor, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor | |
| HEK293 human embryonic kidney cell line transfected with human FPR1, Met1-Lys350, Accession # NP_002020 | |
| 0.1 mL | |
| GPCR, Immunology, Protein Kinase | |
| 2357 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 350418 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction