missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPR1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56473AF350
This item is not returnable.
View return policy
Description
FPR1 Polyclonal antibody specifically detects FPR1 in Human, Mouse, Primate samples. It is validated for Western Blot, Simple Western
Specifications
| FPR1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
| Western Blot, Simple Western | |
| Alexa Fluor 350 | |
| fMet-Leu-Phe receptor, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor | |
| A synthetic peptide corresponding to amino acids 176-190 of human FPR1 (CTFNFSPWTNDPKER) was used as the immunogen | |
| 0.1 mL | |
| GPCR, Immunology, Protein Kinase | |
| 2357 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction