missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRL2/FPR3 Antibody (374822), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3896E
This item is not returnable.
View return policy
Description
FPRL2/FPR3 Monoclonal antibody specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FPRL2/FPR3 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 374822 | |
| Flow Cytometry, CyTOF-ready | |
| FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
| HEK293 human embryonic kidney cell line transfected with human FPRL2, Met1-Met352, Accession # P25089 | |
| 0.1 mL | |
| GPCR, Immunology | |
| 2359 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction